The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286772.1 | NP_001273701.1 | 3 - 4 | 285 | 78 |
NM_018201.4 | NP_060671.3 | 3 - 4 | 285 | 78 | |
GenBank mRNA | AB449899.1 | - | 3 - 4 | 138 | 78 |
AK001605.1 | - | 3 - 4 | 143 | 78 | |
AK289777.1 | - | 3 - 4 | 197 | 78 | |
BC014898.1 | - | 3 - 4 | 160 | 78 | |
BC132817.1 | - | 3 - 4 | 158 | 78 | |
BC136509.1 | - | 3 - 4 | 158 | 78 | |
BC150311.1 | - | 3 - 4 | 194 | 78 | |
DC325060.1 | - | 3 - 4 | 261 | 78 |