The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003355.2 | NP_003346.2 | 3 - 4 | 500 | 66 |
GenBank mRNA | AK025742.1 | - | 3 - 4 | 963 | 66 |
AK222540.1 | - | 3 - 4 | 499 | 66 | |
AK222557.1 | - | 3 - 4 | 499 | 66 | |
BC011737.2 | - | 3 - 4 | 463 | 66 | |
U76367.1 | - | 1 - 2 | 120 | 66 | |
U82819.1 | - | 1 - 2 | 208 | 66 | |
U94592.1 | - | 3 - 4 | 434 | 66 |