The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256695.1 | NP_001243624.1 | 2 - 3 | 371 | 152 |
NM_001256696.1 | NP_001243625.1 | 2 - 3 | 371 | 152 | |
NR_046338.1 | 3 - 4 | 477 | 152 | ||
XM_011520222.2 | XP_011518524.1 | 2 - 3 | 519 | 152 | |
XM_011520223.2 | XP_011518525.1 | 2 - 3 | 201 | 152 | |
XM_011520224.2 | XP_011518526.1 | 2 - 3 | 514 | 152 | |
XM_011520225.2 | XP_011518527.1 | 2 - 3 | 151 | 152 | |
XM_011520228.2 | XP_011518530.1 | 2 - 3 | 512 | 152 | |
XM_011520230.2 | XP_011518532.1 | 2 - 3 | 504 | 152 | |
XM_011520231.2 | XP_011518533.1 | 2 - 3 | 506 | 152 | |
GenBank mRNA | AK094792.1 | - | 2 - 3 | 371 | 152 |