The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128427.2 | NP_001121899.1 | 10 - 11 | 1741 | 97 |
NM_020116.4 | NP_064501.2 | 10 - 11 | 1744 | 97 | |
GenBank mRNA | AB033089.1 | - | 10 - 11 | 1671 | 97 |
AL137695.1 | - | 7 - 8 | 1087 | 97 | |
BC036502.2 | - | 10 - 11 | 1745 | 97 |