Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSTL5
Entrez Gene ID
56884
Gene Name
follistatin like 5
Gene Aliases
-
Chromosome Location
Chr.4: 161383892 - 162164034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128427.2 NP_001121899.1 7 - 8 1326 137
NM_001128428.2 NP_001121900.1 7 - 8 1326 137
NM_020116.4 NP_064501.2 7 - 8 1329 137
XM_011532126.1 XP_011530428.1 7 - 8 1295 137
GenBank mRNA AB033089.1 - 7 - 8 1256 137
AK315152.1 - 7 - 8 1297 137
AL137695.1 - 4 - 5 672 137
BC036502.2 - 7 - 8 1330 137

View Full Product Details