The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128427.2 | NP_001121899.1 | 9 - 10 | 1605 | 109 |
NM_001128428.2 | NP_001121900.1 | 9 - 10 | 1605 | 109 | |
NM_020116.4 | NP_064501.2 | 9 - 10 | 1608 | 109 | |
XM_011532126.1 | XP_011530428.1 | 9 - 10 | 1574 | 109 | |
GenBank mRNA | AB033089.1 | - | 9 - 10 | 1535 | 109 |
AK315152.1 | - | 9 - 10 | 1576 | 109 | |
AL137695.1 | - | 6 - 7 | 951 | 109 | |
BC036502.2 | - | 9 - 10 | 1609 | 109 |