The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005264023.1 | XP_005264080.1 | 4 - 5 | 480 | 145 |
XM_005264024.2 | XP_005264081.1 | 6 - 7 | 645 | 145 | |
XM_005264025.1 | XP_005264082.1 | 5 - 6 | 616 | 145 | |
XM_006712781.2 | XP_006712844.1 | 7 - 8 | 679 | 145 | |
XM_011511934.1 | XP_011510236.1 | 4 - 5 | 515 | 145 | |
XM_011511937.1 | XP_011510239.1 | 4 - 5 | 486 | 145 | |
XM_017005035.1 | XP_016860524.1 | 4 - 5 | 506 | 145 | |
XM_017005036.1 | XP_016860525.1 | 4 - 5 | 515 | 145 | |
XM_017005037.1 | XP_016860526.1 | 4 - 5 | 515 | 145 | |
XM_017005038.1 | XP_016860527.1 | 4 - 5 | 515 | 145 | |
XM_017005039.1 | XP_016860528.1 | 7 - 8 | 668 | 145 | |
XM_017005040.1 | XP_016860529.1 | 6 - 7 | 769 | 145 | |
XM_017005041.1 | XP_016860530.1 | 4 - 5 | 627 | 145 | |
XM_017005042.1 | XP_016860531.1 | 6 - 7 | 657 | 145 | |
XM_017005043.1 | XP_016860532.1 | 4 - 5 | 490 | 145 | |
XM_017005044.1 | XP_016860533.1 | 6 - 7 | 633 | 145 | |
XM_017005047.1 | XP_016860536.1 | 3 - 4 | 543 | 145 | |
GenBank mRNA | AF254756.1 | - | 3 - 4 | 501 | 145 |