Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC6
Entrez Gene ID
368
Gene Name
ATP binding cassette subfamily C member 6
Gene Aliases
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7
Chromosome Location
Chr.16: 16149565 - 16223616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171.5 NP_001162.4 30 - 31 4440 52
XM_011522479.2 XP_011520781.1 30 - 31 4551 52
XM_011522480.1 XP_011520782.1 30 - 31 4388 52
XM_011522481.2 XP_011520783.1 30 - 31 4587 52
XM_017023212.1 XP_016878701.1 29 - 30 4416 52
XM_017023213.1 XP_016878702.1 30 - 31 4474 52
GenBank mRNA AF076622.1 - 30 - 31 4426 52
AF168791.1 - 30 - 31 4403 52
AM711638.1 - 28 - 29 4028 52
AM774324.1 - 30 - 31 4403 52
BC131732.1 - 30 - 31 4426 52

View Full Product Details