The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000830.4 | NP_000821.1 | 9 - 10 | 1805 | 80 |
NM_001320616.1 | NP_001307545.1 | 9 - 10 | 1805 | 80 | |
XM_006723991.1 | XP_006724054.1 | 9 - 10 | 1423 | 80 | |
GenBank mRNA | AK296404.1 | - | 9 - 10 | 1750 | 80 |
L19058.1 | - | 9 - 10 | 1674 | 80 |