The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007086.3 | NP_009017.1 | 1 - 2 | 69 | 69 |
XM_006720012.2 | XP_006720075.1 | 1 - 2 | 67 | 69 | |
GenBank mRNA | BC043349.1 | - | 1 - 2 | 57 | 69 |
BC063041.1 | - | 1 - 2 | 45 | 69 | |
DB082418.1 | - | 1 - 2 | 69 | 69 |