The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014167.4 | NP_054886.2 | 2 - 3 | 510 | 159 |
NR_033192.1 | 2 - 3 | 439 | 159 | ||
GenBank mRNA | AF213377.1 | - | 2 - 3 | 477 | 159 |
BC020647.1 | - | 2 - 3 | 473 | 159 | |
BM846580.1 | - | 2 - 3 | 409 | 159 |