The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014167.4 | NP_054886.2 | 3 - 4 | 606 | 79 |
NR_033192.1 | 3 - 4 | 535 | 79 | ||
GenBank mRNA | AF161477.1 | - | 3 - 4 | 580 | 79 |
AF213377.1 | - | 3 - 4 | 573 | 79 | |
AL832749.1 | - | 1 - 2 | 1626 | 79 | |
BC020647.1 | - | 3 - 4 | 569 | 79 | |
BM846580.1 | - | 3 - 4 | 505 | 79 |