The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017921.3 | NP_060391.2 | 9 - 10 | 1149 | 92 |
XM_011524979.1 | XP_011523281.1 | 9 - 10 | 1144 | 92 | |
XM_011524980.1 | XP_011523282.1 | 9 - 10 | 1144 | 92 | |
XM_011524981.1 | XP_011523283.1 | 9 - 10 | 1144 | 92 | |
XM_011524982.2 | XP_011523284.1 | 9 - 10 | 1144 | 92 | |
XM_017024830.1 | XP_016880319.1 | 5 - 6 | 530 | 92 | |
GenBank mRNA | AB040932.1 | - | 9 - 10 | 1056 | 92 |
AK000664.1 | - | 9 - 10 | 1058 | 92 | |
AK023046.1 | - | 7 - 8 | 785 | 92 | |
AK023272.1 | - | 6 - 7 | 728 | 92 | |
AK294473.1 | - | 8 - 9 | 1026 | 92 | |
AL834300.1 | - | 6 - 7 | 661 | 92 | |
BC025930.1 | - | 9 - 10 | 1090 | 92 |