The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006891.3 | NP_008822.2 | 2 - 3 | 369 | 84 |
GenBank mRNA | BC117338.1 | - | 2 - 3 | 369 | 84 |
BC117340.1 | - | 2 - 3 | 369 | 84 | |
CD674533.1 | - | 1 - 2 | 233 | 84 | |
EL946552.1 | - | 2 - 3 | 285 | 84 | |
KT966392.1 | - | 2 - 3 | 277 | 84 | |
U66583.1 | - | 1 - 2 | 253 | 84 |