The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159936.1 | NP_001153408.1 | 7 - 8 | 921 | 88 |
NM_006824.2 | NP_006815.2 | 6 - 7 | 804 | 88 | |
GenBank mRNA | AB451256.1 | - | 6 - 7 | 607 | 88 |
AB451381.1 | - | 6 - 7 | 607 | 88 | |
AK223455.1 | - | 6 - 7 | 787 | 88 | |
AK299476.1 | - | 4 - 5 | 553 | 88 | |
AK307440.1 | - | 6 - 7 | 750 | 88 | |
BC009175.2 | - | 6 - 7 | 745 | 88 | |
BC073924.1 | - | 12 - 13 | 1156 | 88 | |
BT006920.1 | - | 6 - 7 | 607 | 88 | |
CR456975.1 | - | 6 - 7 | 607 | 88 | |
U86602.1 | - | 6 - 7 | 749 | 88 |