The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199238.1 | NP_001186167.1 | 1 - 2 | 293 | 56 |
NM_001199239.1 | NP_001186168.1 | 1 - 2 | 293 | 56 | |
GenBank mRNA | AK314056.1 | - | 1 - 2 | 293 | 56 |