The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000578.3 | NP_000569.3 | 8 - 9 | 1132 | 63 |
XM_005246793.3 | XP_005246850.1 | 7 - 8 | 1252 | 63 | |
XM_005246794.3 | XP_005246851.1 | 9 - 10 | 1469 | 63 | |
XM_006712709.3 | XP_006712772.1 | 8 - 9 | 1326 | 63 | |
XM_006712710.3 | XP_006712773.1 | 7 - 8 | 1203 | 63 | |
XM_006712711.3 | XP_006712774.1 | 6 - 7 | 1129 | 63 | |
XM_011511684.2 | XP_011509986.1 | 9 - 10 | 1504 | 63 | |
XM_011511685.2 | XP_011509987.1 | 8 - 9 | 1361 | 63 | |
XM_017004765.1 | XP_016860254.1 | 7 - 8 | 1272 | 63 | |
XM_017004766.1 | XP_016860255.1 | 8 - 9 | 1363 | 63 | |
GenBank mRNA | AB209650.1 | - | 6 - 7 | 2846 | 63 |
AK298667.1 | - | 8 - 9 | 886 | 63 | |
AK303398.1 | - | 8 - 9 | 856 | 63 | |
AK307954.1 | - | 8 - 9 | 1072 | 63 | |
AK310914.1 | - | 8 - 9 | 888 | 63 | |
AK311405.1 | - | 7 - 8 | 739 | 63 | |
BC033754.1 | - | 6 - 7 | 1138 | 63 | |
BC071165.1 | - | 9 - 10 | 926 | 63 | |
D38171.1 | - | 6 - 7 | 639 | 63 | |
D50402.1 | - | 8 - 9 | 967 | 63 | |
D50403.1 | - | 8 - 9 | 878 | 63 | |
L32185.1 | - | 8 - 9 | 868 | 63 |