The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144563.2 | NP_653164.2 | 3 - 4 | 449 | 61 |
GenBank mRNA | AY050633.1 | - | 3 - 4 | 425 | 61 |
BC015529.1 | - | 3 - 4 | 421 | 61 | |
CB989690.1 | - | 3 - 4 | 483 | 61 |