The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153332.3 | NP_699163.2 | 3 - 4 | 762 | 138 |
XM_005272401.2 | XP_005272458.1 | 3 - 4 | 617 | 138 | |
XM_011543848.2 | XP_011542150.1 | 3 - 4 | 696 | 138 | |
XM_011543849.1 | XP_011542151.1 | 3 - 4 | 696 | 138 | |
XM_011543850.2 | XP_011542152.1 | 3 - 4 | 801 | 138 | |
XM_011543851.1 | XP_011542153.1 | 4 - 5 | 816 | 138 | |
XM_017013949.1 | XP_016869438.1 | 7 - 8 | 1709 | 138 | |
XM_017013950.1 | XP_016869439.1 | 3 - 4 | 1446 | 138 | |
XM_017013951.1 | XP_016869440.1 | 4 - 5 | 1168 | 138 | |
XM_017013952.1 | XP_016869441.1 | 4 - 5 | 1489 | 138 | |
XM_017013953.1 | XP_016869442.1 | 4 - 5 | 739 | 138 | |
XM_017013954.1 | XP_016869443.1 | 4 - 5 | 1593 | 138 | |
XM_017013955.1 | XP_016869444.1 | 3 - 4 | 813 | 138 | |
GenBank mRNA | AK226180.1 | - | 3 - 4 | 630 | 138 |
AK291062.1 | - | 3 - 4 | 632 | 138 | |
AL137679.1 | - | 3 - 4 | 722 | 138 | |
AY310909.1 | - | 3 - 4 | 626 | 138 | |
BC035279.1 | - | 3 - 4 | 626 | 138 |