Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
Chromosome Location
Chr.19: 3880620 - 3928082 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033064.4 NP_149053.1 10 - 11 1434 76
GenBank mRNA AB058775.1 - 10 - 11 1380 76
AK027889.1 - 10 - 11 1368 76
AK092309.1 - 10 - 11 1233 76
AK125457.1 - 10 - 11 955 76
AK299522.1 - 11 - 12 1440 76
AY220297.1 - 9 - 10 1004 76
BC026217.1 - 10 - 11 1376 76
KU178814.1 - 9 - 10 1004 76

View Full Product Details