The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033064.4 | NP_149053.1 | 5 - 6 | 974 | 74 |
GenBank mRNA | AB058775.1 | - | 5 - 6 | 920 | 74 |
AK027889.1 | - | 5 - 6 | 908 | 74 | |
AK092309.1 | - | 5 - 6 | 773 | 74 | |
AK125457.1 | - | 5 - 6 | 495 | 74 | |
AK299522.1 | - | 6 - 7 | 980 | 74 | |
AY220297.1 | - | 4 - 5 | 544 | 74 | |
BC026217.1 | - | 5 - 6 | 916 | 74 | |
KU178814.1 | - | 4 - 5 | 544 | 74 | |
KU178815.1 | - | 4 - 5 | 544 | 74 |