The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033064.4 | NP_149053.1 | 6 - 7 | 1073 | 68 |
GenBank mRNA | AB058775.1 | - | 6 - 7 | 1019 | 68 |
AK027889.1 | - | 6 - 7 | 1007 | 68 | |
AK092309.1 | - | 6 - 7 | 872 | 68 | |
AK125457.1 | - | 6 - 7 | 594 | 68 | |
AK299522.1 | - | 7 - 8 | 1079 | 68 | |
AY220297.1 | - | 5 - 6 | 643 | 68 | |
BC026217.1 | - | 6 - 7 | 1015 | 68 | |
KU178814.1 | - | 5 - 6 | 643 | 68 | |
KU178815.1 | - | 5 - 6 | 643 | 68 |