Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX3
Entrez Gene ID
8724
Gene Name
sorting nexin 3
Gene Aliases
Grd19, MCOPS8, SDP3
Chromosome Location
Chr.6: 108211217 - 108261260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300928.1 NP_001287857.1 1 - 2 499 102
NM_003795.5 NP_003786.1 1 - 2 499 102
GenBank mRNA AF034546.1 - 1 - 2 203 102
AF062483.1 - 1 - 2 179 102
AK289476.1 - 1 - 2 290 102
BC008444.1 - 1 - 2 257 102
BC014580.1 - 1 - 2 281 102
BC015179.1 - 1 - 2 263 102
BC016863.2 - 1 - 2 513 102
BT007114.1 - 1 - 2 160 102
CR456898.1 - 1 - 2 160 102
HY156437.1 - 1 - 2 337 102

View Full Product Details