Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
Chromosome Location
Chr.8: 138588235 - 138914087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152888.2 NP_690848.1 19 - 20 2406 61
XM_011516883.2 XP_011515185.1 19 - 20 2484 61
XM_011516884.2 XP_011515186.1 18 - 19 2445 61
XM_011516885.2 XP_011515187.1 19 - 20 2484 61
XM_011516886.2 XP_011515188.1 20 - 21 2396 61
XM_011516887.2 XP_011515189.1 16 - 17 1364 61
XM_011516888.2 XP_011515190.1 19 - 20 2487 61
XM_011516889.2 XP_011515191.1 9 - 10 605 61
XM_017013150.1 XP_016868639.1 20 - 21 2357 61
XM_017013151.1 XP_016868640.1 19 - 20 2484 61
XM_017013152.1 XP_016868641.1 15 - 16 1354 61
GenBank mRNA AF406780.1 - 19 - 20 2393 61
BC144535.1 - 8 - 9 1124 61

View Full Product Details