The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152888.2 | NP_690848.1 | 25 - 26 | 2707 | 66 |
XM_011516883.2 | XP_011515185.1 | 25 - 26 | 2785 | 66 | |
XM_011516884.2 | XP_011515186.1 | 24 - 25 | 2746 | 66 | |
XM_011516886.2 | XP_011515188.1 | 26 - 27 | 2697 | 66 | |
XM_011516887.2 | XP_011515189.1 | 22 - 23 | 1665 | 66 | |
XM_011516888.2 | XP_011515190.1 | 25 - 26 | 2788 | 66 | |
XM_011516889.2 | XP_011515191.1 | 15 - 16 | 906 | 66 | |
XM_017013150.1 | XP_016868639.1 | 26 - 27 | 2658 | 66 | |
XM_017013152.1 | XP_016868641.1 | 21 - 22 | 1655 | 66 | |
GenBank mRNA | AF406780.1 | - | 25 - 26 | 2694 | 66 |
BC144535.1 | - | 14 - 15 | 1425 | 66 |