Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT10
Entrez Gene ID
341359
Gene Name
synaptotagmin 10
Gene Aliases
-
Chromosome Location
Chr.12: 33375413 - 33439819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198992.3 NP_945343.1 5 - 6 1669 71
XM_011520644.2 XP_011518946.1 5 - 6 1030 71
GenBank mRNA AY198413.1 - 5 - 6 1670 71
BC101024.2 - 5 - 6 1496 71
BC101025.1 - 7 - 8 71

View Full Product Details