Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK000394.1 - 1 - 2 1468 Chr. 1 - 94621137 - 94624184 [-] on Build GRCh38 79


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream coagulation factor III, tissue factor F3 CD142, TF, TFA -79.3kb Sense Chr. - 94529176 - 94541857 on Build GRCh38
Downstream long intergenic non-protein coding RNA 1057 LINC01057 XLOC_000303 33.3kb Sense Chr. - 94657533 - 94820281 on Build GRCh38

View Full Product Details