The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_036632.1 | 8 - 9 | 1573 | 106 | |
GenBank mRNA | AF217995.1 | - | 8 - 9 | 1523 | 106 |
BC109389.1 | - | 4 - 5 | 608 | 106 | |
BC110077.1 | - | 4 - 5 | 608 | 106 | |
BC141930.1 | - | 4 - 5 | 608 | 106 |