Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PYROXD2
Entrez Gene ID
84795
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Gene Aliases
C10orf33, FP3420
Chromosome Location
Chr.10: 98383565 - 98415221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032709.2 NP_116098.2 11 - 12 1218 50
XM_011540293.2 XP_011538595.1 12 - 13 2640 50
XM_011540301.2 XP_011538603.1 4 - 5 2054 50
XM_017016835.1 XP_016872324.1 11 - 12 1179 50
XM_017016836.1 XP_016872325.1 11 - 12 1179 50
XM_017016837.1 XP_016872326.1 11 - 12 1179 50
XM_017016838.1 XP_016872327.1 11 - 12 3414 50
XM_017016839.1 XP_016872328.1 9 - 10 1271 50
XM_017016840.1 XP_016872329.1 9 - 10 1125 50
XM_017016841.1 XP_016872330.1 8 - 9 1432 50
GenBank mRNA AK074429.1 - 11 - 12 1179 50
AK075265.1 - 11 - 12 1156 50
AK123713.1 - 1 - 2 50
AK225637.1 - 11 - 12 1172 50
BC006131.1 - 11 - 12 1181 50

View Full Product Details