The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321073.1 | NP_001308002.1 | 2 - 3 | 244 | 118 |
XM_011546094.1 | XP_011544396.1 | 2 - 3 | 136 | 118 | |
XM_011546095.2 | XP_011544397.1 | 2 - 3 | 278 | 118 | |
XM_011546097.2 | XP_011544399.1 | 3 - 4 | 305 | 118 | |
XM_011546098.1 | XP_011544400.1 | 2 - 3 | 643 | 118 | |
XM_011546099.1 | XP_011544401.1 | 2 - 3 | 136 | 118 | |
XM_017028211.1 | XP_016883700.1 | 3 - 4 | 641 | 118 | |
XM_017028212.1 | XP_016883701.1 | 4 - 5 | 697 | 118 | |
XM_017028213.1 | XP_016883702.1 | 2 - 3 | 136 | 118 | |
XM_017028214.1 | XP_016883703.1 | 3 - 4 | 638 | 118 | |
XM_017028215.1 | XP_016883704.1 | 3 - 4 | 253 | 118 | |
XM_017028216.1 | XP_016883705.1 | 2 - 3 | 643 | 118 | |
XM_017028217.1 | XP_016883706.1 | 2 - 3 | 136 | 118 | |
XM_017028218.1 | XP_016883707.1 | 3 - 4 | 305 | 118 | |
GenBank mRNA | BC007257.1 | - | 2 - 3 | 214 | 118 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000071.2 | NP_000062.1 | 2 - 3 | 237 | 118 |
NM_001178008.2 | NP_001171479.1 | 2 - 3 | 244 | 118 | |
NM_001178009.2 | NP_001171480.1 | 2 - 3 | 244 | 118 | |
NM_001320298.1 | NP_001307227.1 | 3 - 4 | 238 | 118 | |
XM_011529777.1 | XP_011528079.1 | 2 - 3 | 180 | 118 | |
XM_011529781.1 | XP_011528083.1 | 3 - 4 | 245 | 118 | |
XM_011529782.1 | XP_011528084.1 | 2 - 3 | 643 | 118 | |
XM_017028489.1 | XP_016883978.1 | 3 - 4 | 640 | 118 | |
XM_017028490.1 | XP_016883979.1 | 3 - 4 | 637 | 118 | |
XM_017028491.1 | XP_016883980.1 | 2 - 3 | 180 | 118 | |
XM_017028492.1 | XP_016883981.1 | 3 - 4 | 245 | 118 | |
GenBank mRNA | AK313691.1 | - | 2 - 3 | 142 | 118 |
AL531222.3 | - | 2 - 3 | 228 | 118 | |
BC000440.2 | - | 2 - 3 | 112 | 118 | |
BC010242.1 | - | 2 - 3 | 161 | 118 | |
BC011381.1 | - | 2 - 3 | 100 | 118 | |
DC300040.1 | - | 2 - 3 | 237 | 118 | |
DC315340.1 | - | 3 - 4 | 238 | 118 | |
HY151461.1 | - | 2 - 3 | 245 | 118 | |
L19501.1 | - | 2 - 3 | 173 | 118 | |
X82166.1 | - | 2 - 3 | 139 | 118 |