The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099676.2 | NP_001093146.1 | 5 - 6 | 740 | 100 |
NM_001170633.1 | NP_001164104.1 | 7 - 8 | 1220 | 100 | |
XM_017018770.1 | XP_016874259.1 | 7 - 8 | 1235 | 100 | |
GenBank mRNA | BC014352.1 | - | 2 - 3 | 211 | 100 |
BC015121.1 | - | 2 - 3 | 211 | 100 | |
BC039369.1 | - | 5 - 6 | 688 | 100 | |
BX089271.1 | - | 2 - 3 | 178 | 100 |