The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182521.2 | NP_872327.2 | 6 - 7 | 866 | 155 |
GenBank mRNA | AK128006.1 | - | 6 - 7 | 866 | 155 |
BC031094.1 | - | 6 - 7 | 865 | 155 |