Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFS1
Entrez Gene ID
4719
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
Gene Aliases
CI-75Kd, CI-75k, PRO1304
Chromosome Location
Chr.2: 206121971 - 206159519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199981.1 NP_001186910.1 4 - 5 416 105
NM_001199983.1 NP_001186912.1 4 - 5 432 105
NM_001199984.1 NP_001186913.1 5 - 6 444 105
NM_005006.6 NP_004997.4 5 - 6 524 105
XM_017004188.1 XP_016859677.1 5 - 6 452 105
GenBank mRNA AK295705.1 - 4 - 5 322 105
AK295987.1 - 5 - 6 444 105
AK300585.1 - 4 - 5 336 105
BC018900.1 - 5 - 6 400 105
BC022368.1 - 5 - 6 425 105
BC030833.1 - 5 - 6 428 105
BQ882763.1 - 5 - 6 468 105

View Full Product Details