Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPT2
Entrez Gene ID
4735
Gene Name
septin 2
Gene Aliases
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
Chromosome Location
Chr.2: 241315187 - 241354027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008491.2 NP_001008491.1 9 - 10 1244 157
NM_001008492.2 NP_001008492.1 9 - 10 926 157
NM_001282972.1 NP_001269901.1 9 - 10 975 157
NM_001282973.1 NP_001269902.1 12 - 13 1199 157
NM_001321029.1 NP_001307958.1 9 - 10 960 157
NM_001321030.1 NP_001307959.1 9 - 10 958 157
NM_001321031.1 NP_001307960.1 10 - 11 1061 157
NM_001321032.1 NP_001307961.1 8 - 9 841 157
NM_001321033.1 NP_001307962.1 10 - 11 1077 157
NM_001321034.1 NP_001307963.1 10 - 11 1076 157
NM_001321035.1 NP_001307964.1 9 - 10 942 157
NM_004404.4 NP_004395.1 8 - 9 841 157
NM_006155.2 NP_006146.1 8 - 9 1143 157
XM_011511237.1 XP_011509539.1 8 - 9 807 157
XM_011511238.1 XP_011509540.1 7 - 8 675 157
XM_011511240.1 XP_011509542.1 7 - 8 684 157
XM_017004189.1 XP_016859678.1 10 - 11 1036 157
XM_017004190.1 XP_016859679.1 10 - 11 971 157
XM_017004191.1 XP_016859680.1 10 - 11 1036 157
XM_017004192.1 XP_016859681.1 10 - 11 1036 157
XM_017004193.1 XP_016859682.1 8 - 9 841 157
XM_017004194.1 XP_016859683.1 10 - 11 991 157
XM_017004195.1 XP_016859684.1 10 - 11 1195 157
XM_017004196.1 XP_016859685.1 10 - 11 1077 157
XM_017004197.1 XP_016859686.1 9 - 10 892 157
XM_017004198.1 XP_016859687.1 9 - 10 954 157
XM_017004199.1 XP_016859688.1 10 - 11 993 157
XM_017004200.1 XP_016859689.1 9 - 10 956 157
XM_017004201.1 XP_016859690.1 9 - 10 1078 157
XM_017004202.1 XP_016859691.1 8 - 9 1060 157
XM_017004203.1 XP_016859692.1 9 - 10 884 157
XM_017004204.1 XP_016859693.1 8 - 9 799 157
XM_017004205.1 XP_016859694.1 10 - 11 991 157
XM_017004206.1 XP_016859695.1 9 - 10 892 157
XM_017004207.1 XP_016859696.1 6 - 7 1290 157
XM_017004208.1 XP_016859697.1 9 - 10 1500 157
XM_017004209.1 XP_016859698.1 8 - 9 1387 157
XM_017004210.1 XP_016859699.1 8 - 9 1431 157
XM_017004211.1 XP_016859700.1 7 - 8 1284 157
XM_017004212.1 XP_016859701.1 7 - 8 1536 157
GenBank mRNA AF038404.1 - 8 - 9 780 157
AK294563.1 - 9 - 10 883 157
BC014455.1 - 9 - 10 1123 157
BC033559.1 - 12 - 13 1177 157
BX648000.1 - 9 - 10 860 157
D28540.1 - 7 - 8 764 157
D63878.1 - 8 - 9 960 157

View Full Product Details