Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF035314.1 - 1 685 Chr. 20 - 21238182 - 21240177 [+] on Build GRCh38 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream KIZ antisense RNA 1 KIZ-AS1 -19.9kb Anti-sense Chr. - 21194404 - 21218289 on Build GRCh38
Downstream 5'-3' exoribonuclease 2 XRN2 63.1kb Sense Chr. - 21303284 - 21389825 on Build GRCh38

View Full Product Details