Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPC2
Entrez Gene ID
221914
Gene Name
glypican 2
Gene Aliases
-
Chromosome Location
Chr.7: 100169606 - 100177426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152742.2 NP_689955.1 3 - 4 871 119
GenBank mRNA AK096281.1 - 3 - 4 809 119
AL834418.1 - 3 - 4 757 119
AY093588.1 - 3 - 4 818 119
BC027972.1 - 3 - 4 812 119

View Full Product Details