Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC36A3
Entrez Gene ID
285641
Gene Name
solute carrier family 36 member 3
Gene Aliases
PAT3, TRAMD2, tramdorin2
Chromosome Location
Chr.5: 151276365 - 151303773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181774.3 NP_861439.3 4 - 5 828 62
XM_011537627.2 XP_011535929.1 4 - 5 828 62
XM_011537629.2 XP_011535931.1 4 - 5 828 62
XM_011537630.1 XP_011535932.1 2 - 3 175 62
XM_011537631.1 XP_011535933.1 2 - 3 186 62
XM_011537634.2 XP_011535936.1 4 - 5 828 62
GenBank mRNA AK127978.1 - 4 - 5 821 62
AY162215.1 - 4 - 5 646 62
BC101094.1 - 4 - 5 551 62
BC101095.2 - 4 - 5 551 62

View Full Product Details