The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145017.1 | NP_001138489.1 | 7 - 8 | 1242 | 91 |
NM_181774.3 | NP_861439.3 | 6 - 7 | 1119 | 91 | |
XM_011537629.2 | XP_011535931.1 | 6 - 7 | 1119 | 91 | |
XM_011537630.1 | XP_011535932.1 | 4 - 5 | 466 | 91 | |
XM_011537631.1 | XP_011535933.1 | 4 - 5 | 477 | 91 | |
XM_011537633.1 | XP_011535935.1 | 2 - 3 | 329 | 91 | |
GenBank mRNA | AK127978.1 | - | 6 - 7 | 1112 | 91 |
AK131483.1 | - | 5 - 6 | 2600 | 91 | |
AY162215.1 | - | 6 - 7 | 937 | 91 | |
BC101092.1 | - | 7 - 8 | 965 | 91 | |
BC101093.1 | - | 7 - 8 | 965 | 91 | |
BC101094.1 | - | 6 - 7 | 842 | 91 | |
BC101095.2 | - | 6 - 7 | 842 | 91 |