The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199397.1 | NP_001186326.1 | 1 - 2 | 402 | 139 |
NM_001199398.1 | NP_001186327.1 | 1 - 2 | 402 | 139 | |
NM_001199399.1 | NP_001186328.1 | 1 - 2 | 402 | 139 | |
XM_006714228.1 | XP_006714291.1 | 1 - 2 | 404 | 139 | |
XM_011532003.1 | XP_011530305.1 | 1 - 2 | 404 | 139 | |
XM_011532004.1 | XP_011530306.1 | 1 - 2 | 404 | 139 | |
XM_011532005.2 | XP_011530307.1 | 1 - 2 | 404 | 139 | |
XM_017008249.1 | XP_016863738.1 | 1 - 2 | 404 | 139 | |
XM_017008251.1 | XP_016863740.1 | 1 - 2 | 404 | 139 | |
XM_017008253.1 | XP_016863742.1 | 1 - 2 | 404 | 139 | |
GenBank mRNA | AL050385.1 | - | 1 - 2 | 357 | 139 |
BC068201.1 | - | 1 - 2 | 362 | 139 | |
BC114491.1 | - | 1 - 2 | 162 | 139 | |
CR933642.1 | - | 1 - 2 | 357 | 139 | |
DA697159.1 | - | 1 - 2 | 403 | 139 |