Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM43
Entrez Gene ID
375287
Gene Name
RNA binding motif protein 43
Gene Aliases
C2orf38
Chromosome Location
Chr.2: 151247689 - 151261875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198557.2 NP_940959.1 3 - 4 459 141
XM_017004052.1 XP_016859541.1 3 - 4 916 141
GenBank mRNA AK127552.1 - 3 - 4 448 141
BC136410.1 - 3 - 4 359 141
BC136411.1 - 3 - 4 359 141
BX337439.2 - 3 - 4 465 141
DA658209.1 - 3 - 4 437 141

View Full Product Details