The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308036.1 | NP_001294965.1 | 2 - 3 | 685 | 89 |
NM_007362.3 | NP_031388.2 | 2 - 3 | 347 | 89 | |
XM_011512556.2 | XP_011510858.1 | 2 - 3 | 404 | 89 | |
XM_011512557.2 | XP_011510859.1 | 2 - 3 | 452 | 89 | |
XM_011512558.2 | XP_011510860.1 | 2 - 3 | 294 | 89 | |
GenBank mRNA | AK093216.1 | - | 2 - 3 | 397 | 89 |
AK296144.1 | - | 2 - 3 | 311 | 89 | |
AK297506.1 | - | 2 - 3 | 650 | 89 | |
AK308192.1 | - | 2 - 3 | 351 | 89 | |
AK315903.1 | - | 2 - 3 | 321 | 89 | |
AK316601.1 | - | 2 - 3 | 314 | 89 | |
AY644767.1 | - | 2 - 3 | 271 | 89 | |
BC001255.1 | - | 2 - 3 | 283 | 89 | |
BP248419.1 | - | 2 - 3 | 347 | 89 | |
BT006842.1 | - | 2 - 3 | 257 | 89 | |
BX647994.1 | - | 2 - 3 | 332 | 89 | |
X84157.1 | - | 2 - 3 | 286 | 89 |