Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYLS1
Entrez Gene ID
219844
Gene Name
HYLS1, centriolar and ciliogenesis associated
Gene Aliases
HLS
Chromosome Location
Chr.11: 125883614 - 125900646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134793.1 NP_001128265.1 3 - 3 1994 84
NM_145014.2 NP_659451.1 4 - 4 1747 84
XM_005271430.2 XP_005271487.1 2 - 2 1944 84
XM_006718777.3 XP_006718840.1 2 - 2 1261 84
XM_011542659.2 XP_011540961.1 2 - 2 1422 84
XM_017017320.1 XP_016872809.1 3 - 3 1713 84
XM_017017321.1 XP_016872810.1 3 - 3 1710 84
GenBank mRNA AK057477.1 - 4 - 4 1749 84
AK127394.1 - 3 - 3 1997 84
BC015047.1 - 3 - 3 1335 84
BM970011.1 - 1 - 1 84

View Full Product Details