Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGFL7
Entrez Gene ID
51162
Gene Name
EGF like domain multiple 7
Gene Aliases
NEU1, VE-STATIN, ZNEU1
Chromosome Location
Chr.9: 136654753 - 136672678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016215.4 NP_057299.1 11 - 11 1684 98
NM_201446.2 NP_958854.1 9 - 9 1238 98
NR_045110.1 10 - 10 1465 98
NR_045111.1 10 - 10 1468 98
NR_046367.1 10 - 10 2061 98
XM_006717141.3 XP_006717204.1 8 - 8 1065 98
XM_011518764.1 XP_011517066.1 11 - 11 2385 98
XM_011518765.1 XP_011517067.1 11 - 11 2382 98
XM_011518766.1 XP_011517068.1 12 - 12 2601 98
XM_011518767.1 XP_011517069.1 8 - 8 1823 98
XM_011518768.1 XP_011517070.1 8 - 8 1820 98
XM_017014795.1 XP_016870284.1 9 - 9 2147 98
XM_017014796.1 XP_016870285.1 9 - 9 2144 98
XM_017014797.1 XP_016870286.1 10 - 10 2223 98
XM_017014798.1 XP_016870287.1 10 - 10 2360 98
XM_017014799.1 XP_016870288.1 10 - 10 2363 98
GenBank mRNA AF186111.1 - 8 - 8 1169 98
AK091964.1 - 10 - 10 2061 98
AL512735.1 - 10 - 10 1465 98
AY358902.1 - 11 - 11 1637 98
AY358903.1 - 10 - 10 1475 98
BC012377.1 - 9 - 9 1218 98
BC088371.1 - 10 - 10 1458 98
BX647866.1 - 3 - 3 852 98
CB047042.1 - 1 - 1 363 98

View Full Product Details