Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORA16B
Entrez Gene ID
692157
Gene Name
small nucleolar RNA, H/ACA box 16B
Gene Aliases
U98b
Chromosome Location
Chr.1: 212352818 - 212352950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_004389.1 1 - 1 58 86


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CU452616.1 - 1 162 Chr. 1 - 212352718 - 212353356 [+] on Build GRCh38 86
GenBankmRNA CU457294.1 - 1 162 Chr. 1 - 212352718 - 212353484 [+] on Build GRCh38 86
GenBankmRNA CU446454.1 - 1 162 Chr. 1 - 212352718 - 212353455 [+] on Build GRCh38 86
GenBankmRNA CU444851.1 - 1 162 Chr. 1 - 212352718 - 212353442 [+] on Build GRCh38 86
GenBankmRNA CU450146.1 - 1 162 Chr. 1 - 212352718 - 212353394 [+] on Build GRCh38 86
GenBankmRNA CU455673.1 - 1 162 Chr. 1 - 212352718 - 212353428 [+] on Build GRCh38 86


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101929541 LOC101929541 -67.8kb Anti-sense Chr. - 212213002 - 212285021 on Build GRCh38
Downstream transmembrane protein 206 TMEM206 C1orf75 11.5kb Anti-sense Chr. - 212364474 - 212415508 on Build GRCh38

View Full Product Details