The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026732.1 | 2 - 3 | 224 | 81 | |
GenBank mRNA | BM839073.1 | - | 2 - 3 | 143 | 81 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | forkhead box G1 | FOXG1 | BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN | -3.1kb | Sense | Chr. - 28767072 - 28770277 on Build GRCh38 |
Downstream | uncharacterized LOC105370424 | LOC105370424 | 5.5kb | Anti-sense | Chr. - 28783491 - 28785787 on Build GRCh38 |