Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHOBTB1
Entrez Gene ID
9886
Gene Name
Rho related BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 60869438 - 61001486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024554.2 2 - 3 175 100
NR_024555.1 2 - 3 139 100
XM_011540424.1 XP_011538726.1 2 - 3 167 100
XM_011540425.1 XP_011538727.1 2 - 3 167 100
XM_011540426.1 XP_011538728.1 2 - 3 167 100
XM_011540428.1 XP_011538730.1 2 - 3 167 100
XM_017017003.1 XP_016872492.1 2 - 3 185 100
XM_017017004.1 XP_016872493.1 2 - 3 187 100
GenBank mRNA AK075129.1 - 2 - 3 139 100
BC093035.1 - 2 - 3 139 100
CN286750.1 - 2 - 3 128 100
DA781491.1 - 2 - 3 167 100
DR004641.1 - 2 - 3 182 100


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA858646.1 - 2 - 3 140 Chr. 10 - 60892924 - 61001441 [-] on Build GRCh38 100
GenBankmRNA DA830040.1 - 139 Chr. - - [-] on Build 100
GenBankmRNA CB995453.1 - 2 - 3 166 Chr. 10 - 60910735 - 61001440 [-] on Build GRCh38 100
GenBankmRNA BX397555.2 - 1 - 2 102 Chr. 10 - 60888841 - 60985950 [-] on Build GRCh38 100
GenBankmRNA BX336049.2 - 2 - 3 128 Chr. 10 - 60888996 - 61001429 [-] on Build GRCh38 100
GenBankmRNA W88707.1 - 168 Chr. - - [-] on Build 100
GenBankmRNA DA416039.1 - 2 - 3 129 Chr. 10 - 60892922 - 61001430 [-] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cyclin dependent kinase 1 CDK1 CDC2, CDC28A, P34CDC2 -147.0kb Anti-sense Chr. - 60778331 - 60794852 on Build GRCh38
Downstream long intergenic non-protein coding RNA 845 LINC00845 30.3kb Anti-sense Chr. - 61016275 - 61026417 on Build GRCh38

View Full Product Details