Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMNL1
Entrez Gene ID
752
Gene Name
formin like 1
Gene Aliases
C17orf1, C17orf1B, FHOD4, FMNL, KW-13
Chromosome Location
Chr.17: 45221728 - 45247322 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ008116.1 - 1 - 1 52 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC339192 LOC339192 -23.3kb Anti-sense Chr. - 45219313 - 45219974 on Build GRCh38
Downstream spermatogenesis associated 32 SPATA32 AEP2, C17orf46, TEX34, VAD1.2 3.9kb Anti-sense Chr. - 45254393 - 45262112 on Build GRCh38

View Full Product Details