Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NTNG1
- Entrez Gene ID
-
22854
- Gene Name
- netrin G1
- Gene Aliases
-
Lmnt1
- Chromosome Location
-
Chr.1: 107139918 - 107484899 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI050024.1 |
- |
1 |
|
|
Chr. 1 - 107484529 - 107484899 [-] on Build GRCh38 |
92 |
GenBankmRNA |
AI733615.1 |
- |
1 |
|
|
Chr. 1 - 107484400 - 107484899 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protein arginine methyltransferase 6 |
PRMT6 |
HRMT1L6 |
-423.8kb
|
Sense |
Chr. - 107056645 - 107059294 on Build GRCh38 |
Downstream |
vav guanine nucleotide exchange factor 3 |
VAV3 |
|
86.3kb
|
Anti-sense |
Chr. - 107571160 - 107964923 on Build GRCh38 |
View Full Product Details