Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- SNHG6
- Entrez Gene ID
-
641638
- Gene Name
- small nucleolar RNA host gene 6
- Gene Aliases
-
HBII-276HG, NCRNA00058, U87HG
- Chromosome Location
-
Chr.8: 66921930 - 66925542 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057116.1 |
- |
1 - 3 |
2478 |
101 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB332405.1 |
- |
1 - 3 |
|
|
Chr. 8 - 66921995 - 66923465 [+] on Build GRCh38 |
101 |
View Full Product Details