Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI769287.1 |
- |
1 |
|
|
Chr. 8 - 8873482 - 8873858 [+] on Build GRCh38 |
103 |
GenBankmRNA |
AL832142.1 |
- |
|
7145 |
|
Chr. - - [-] on Build |
103 |
GenBankmRNA |
BU753003.1 |
- |
1 |
|
|
Chr. 8 - 8873476 - 8873891 [+] on Build GRCh38 |
103 |
GenBankmRNA |
BQ022625.1 |
- |
1 |
|
|
Chr. 8 - 8873476 - 8873827 [+] on Build GRCh38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
claudin 23 |
CLDN23 |
CLDNL, hCG1646163 |
-169.4kb
|
Sense |
Chr. - 8702156 - 8704107 on Build GRCh38 |
Downstream |
exoribonuclease 1 |
ERI1 |
3'HEXO, HEXO, THEX1 |
128.9kb
|
Sense |
Chr. - 8892225 - 9100262 on Build GRCh38 |
View Full Product Details